Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR epT.I.P.S. Relo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z640506960EA
Encompass
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays with plastic lids protecting the trays from contact during transfer to the working box. Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray or loaded into ep T.I.P.S box (purchase separately). PCR clean free of human DNA DNase RNase and PCR inhibitors batch related certificates available on the internet at http//www.eppendorf.com
Catalog No. 50-290-0685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.