Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Cell Fractionation Buffer for PrepSEQ™ Nucleic Acid Sample Preparation Kit

Catalog No. 4403461
Change view
Click to view available options
Quantity:
3 bottles, 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-034-61 3 bottles, 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-034-61 Supplier Applied Biosystems™ Supplier No. 4403461
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell Fractionation Buffer is used to process the mammalian cell pellet to obtain free Mycoplasma and then combine with the Mycoplasma pellet in the large scale PrepSEQ extraction protocol.

Cell Fractionation Buffer is used to process the mammalian cell pellet to obtain free Mycoplasma and then combine with the Mycoplasma pellet in the large scale PrepSEQ extraction protocol. This protocol can process up to 15 mL of sample (up to 106 cells/mL) for the detection of Mycoplasma.

Specifications

Chemical Name or Material Cell Fractionation Buffer
For Use With (Application) Mycoplasma detection
For Use With (Equipment) PrepSEQ™ Nucleic Acid Sample Preparation Kit
Physical Form Liquid
Quantity 3 bottles, 25 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.