Learn More
Abcam Cell Invasion Assay, Basement Membrane, 96-Well, 8 μm, 96 Tests
Description
- Boyden chamber assay for cell invasion
- Designed for use with suspension and adherent cells
- Detection method: Fluorescent
- Format: 96-well, 8 μm
- Citation count: 12
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate | Basement membrane |
| Target Molecule | Cells |
| Pore Size | 8 μm |
| No. of Reactions | 96 Tests |
| Format | 96-well Plate Kit |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 96 Tests |
| For Use With (Application) | Cell Invasion |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.