Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cell Invasion Assay Kit, Collagen I, 24-Well, 8 μm, 12 Test

Catalog No. 502586486
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6486 12 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6486 Supplier Abcam Supplier No. AB23588712TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fluorescent assay kit for monitoring cell invasion in suspension and adherent cells using a microplate reader.

  • Detection method: Fluorescent
  • Sample types: Suspension cells, Adherent cells
  • Suitable for use with: Microplate reader
  • Citation count: 3

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Format 24-well Plate Kit
Detection Method Fluorescence
Target Molecule Collagen I
Pore Size 8 μm
Substrate Properties Protein-based Substrate
For Use With (Equipment) Microplate Reader
No. of Reactions 12 Tests
Sample Type Suspension cells, Adherent cells
Quantity 12 Tests
For Use With (Application) Cell Invasion Assay, Functional Studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.