Learn More
Aat Bioquest Cell Navigator Cell Plasma Membrane Staining Kit Red Fluorescence (500 Tests)
SDP
Supplier:  Aat Bioquest 22681
Encompass_Preferred
The Cell Navigator Plasma Membrane Stain Kit provides a fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins. This catalog is for 500 Tests.
Catalog No. 50-225-0110