Learn More
Cancer Diagnostics, Inc.™ Cell Safe Biopsy Capsule
SDP
Catalog No. p-3188181 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-500-464 Blue
22-500-463 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-500-464 Supplier Cancer Diagnostics, Inc.™ Supplier No. CB7777
Add to Cart
Add to Cart
Fits inside any cassette
  • Turns any cassette into a mesh cassette—eliminates artifact
  • Disposable plastic biopsy insert consists of two hinged interlocking frames with an integral nylon mesh
  • Extra fine mesh ensures a high level of specimen security
Order Info
Dimensions: 28L x 25W x 5mmH with 18L x 18W x 2mmH mesh area.
Specifications
Color Blue