Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X)
Catalog No. 509305
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-930-5 100 μL
50-112-9036 4 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-930-5 Supplier Invitrogen™ Supplier No. 00497093
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart
Cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines.
  • Can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays
  • Can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
Volume (Metric) 100μL
Product Line eBioscience
Product Type Cell Stimulation Cocktail
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.