Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
Catalog No. 509306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-930-6 4 x 100 μL
50-930-7 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-930-6 Supplier Invitrogen™ Supplier No. 00497503
Only null left
Add to Cart
Add to Cart
Can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
Cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • A cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin
  • Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines
  • Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • Thus, it can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4 x 100 μL
For Use With (Application) Flow Cytometry, Functional Assays, Cell Culture
Product Line eBioscience
Product Type Cell Stimulation Cocktail
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.