Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
Catalog No. 509307
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-930-7 100 μL
50-930-6 4 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-930-7 Supplier Invitrogen™ Supplier No. 00497593
Add to Cart
Edge
Add to Cart
Can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
Cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • A cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin
  • Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines
  • Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
  • Thus, it can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
For Use With (Application) Flow Cytometry, Functional Assays, Cell Culture
Product Line eBioscience
Product Type Cell Stimulation Cocktail
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.