Learn More
STEMCELL Technologies CellAdhere™ Vitronectin, Human, Solution, Size: 0.2 mL
SDP
Supplier:  STEMCELL Technologies 07004
Encompass_Preferred
Vitronectin is a common glycoprotein that is abundant in animal serum, and can also be found in the extracellular matrix deposited by many cell types. It is primarily used in cell culture as a surface coating to promote cell attachment and spreading.CellAdhere™ Vitronectin, Human, Solution is purified from human serum by the method of Hayman et al. using an anti-Vitronectin monoclonal antibody affinity column and sterilized by 0.2 µm filtration. Vitronectin has a purity of > 95% based on Coomassie brilliant blue stain of 7.5% SDS-PAGE. Product is dissolved in 0.2 mL of 0.15 M NaCl, 0.005 M HEPES buffer at approximately pH 7.4. Final concentration of Vitronectin is 0.5 mg/mL. Fibronectin contamination is less than 0.04% based on immunoblotting. The optimal protein concentration may vary depending on the cell type being used, and therefore must be titrated for best results.
Catalog No. 50-197-4382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.