Learn More
Cytiva CELLBAG, 10L, BC10, SCREW, DO
Catalog No. 501055442 Shop All Cytiva Products
Encompass
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.
Catalog No. 50-105-5442 Supplier Cytiva Supplier No. CB0010L1003
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.