Learn More
Cytiva CELLBAG, 2L, BC10, BASIC
Catalog No. 501055437 Shop All Cytiva Products
Encompass
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.
Catalog No. 50-105-5437 Supplier Cytiva Supplier No. CB0002L1001
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.