Learn More
Cytiva CELLBAG, 50L, BC10, BASIC
Catalog No. 501055450 Shop All Cytiva Products
Encompass
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.
Catalog No. 50-105-5450 Supplier Cytiva Supplier No. CB0050L1001
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.