Learn More
Cytiva CELLBAG, 50L, BC10, DO
Catalog No. 501055451 Shop All Cytiva Products
Encompass
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.
Catalog No. 50-105-5451 Supplier Cytiva Supplier No. CB0050L1002
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.