Learn More
Greiner Bio-One CELLCOAT™ 384-well, Collagen Type I-Treated, Flat-Bottom Microplate
Shop All Greiner Bio-One Products

Description
- Promotes growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
- Cell cultivation in serum-free or serum-reduced medium
- Rat tail collagen type I
- Increase in isolation and cu
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 15 to 110 μL |
| Sterility | Non-sterile |
| Surface Treatment | Collagen Type I |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.