Learn More
Greiner Bio-One CELLCOAT™ 96-well, Collagen Type I-Treated, Flat-Bottom Microplate
Shop All Greiner Bio-One Products

Description
- Cell cultivation in serum-free or serum-reduced medium
- Rat tail Collagen Type I
- Increase in isolation and cultivation efficiency
- Immediate use, time-saving
- Consistent quality
- Storable at room temperature
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 25 to 340 μL |
| Sterility | Non-sterile |
| Surface Treatment | Collagen Type I |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.