Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks
GREENER_CHOICE
Catalog No. 07000664 Shop All Greiner Bio-One Products
Change view
Click to view available options
Culture Area:
175 cm2
25 cm2
Volume (Metric) Working:
50 mL
650mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Volume (Metric) Working
07-000-664 175 cm2 650mL
07-000-765 25 cm2 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-664 Supplier Greiner Bio-One Supplier No. 661940
Add to Cart
Add to Cart
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks helps in promotion of cell adhesion and proliferation
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
  • Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
  • For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
  • Ready-to-use products
  • Includes lid with condensation rings
TRUSTED_SUSTAINABILITY
Specifications
Surface Coating Poly-D-Lysine
Closure Type Plug Seal Cap
Culture Area 175 cm2
Volume (Metric) Working 650mL