Learn More
Bulldog Bio Inc CellCover Biomolecule Stabilizing Solution, 50ml bottle
SDP
Supplier:  Bulldog Bio Inc 800050
Encompass_Preferred
For parallel storage of proteins, RNA, and DNA in their cellular contextCellCover was developed for fast “one step” stabilization of biomolecules. Its non-toxic formulation instantaneously protects and preserves RNA and protein expression patterns in human and animal cell types. This includes cells found in solid tissues, such as tumors, and cultured cell clusters, such as organoids and spheroids. Labs can then analyze the treated cell’s DNA, RNA and proteins days or weeks later with almost no detectable change in levels, patterns or modifications.The rapid fixation of RNA and proteins is very important for expression patterns to remain unchanged. With other techniques, such rapid fixation is often at the expense of other factors such as molecule crosslinking by PFA (paraformaldehyde) or cell lysis by high-salt reagents that stabilize RNA. Not so with CellCover, which instantly fixes and stabilizes cells in their true state — maintaining RNA and protein expression.
Catalog No. NC1933412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.