Learn More
Invitrogen™ Celleste™ 2D Deconvolution with GPU Acceleration Module
Catalog No. 12563699
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-699 1 each
1 options
Catalog No. 12-563-699 Supplier Invitrogen™ Supplier No. AMEP4939
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.
This Celleste 2D Deconvolution GPU Acceleration Module single-user license helps deconvolve 2D image sets in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC or workstation.
This Celleste 2D Deconvolution GPU Acceleration Module single-user license helps deconvolve 2D image sets in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC or workstation. It is used in conjunction with Celleste software (Cat. No. AMEP4942) and 2D deconvolution module (AMEP4864) to effectively clear background from images and improve signal-to-noise ratios. A network license including one seat is also available (AMEP4940).
Celleste 6 Image Analysis Software with the 2D Deconvolution and 2D Deconvolution GPU Acceleration modules offers these important advantages:
  • Icon-based wizard-driven workflow streamlines image analysis and eliminates guesswork
  • Preconfigured and optimized analysis templates for common applications help derive the most relevant results from image sets
  • Wide range of report formats to visualize results, create reports, and share images and data
  • Improve signal-to-noise ratios by deconvolution and generate higher quality images and data
  • GPU-accelerated deconvolution to increase performance and reduce the time to results
  • Clear images in a fraction of the time with CPU-based processing
More about deconvolution
Deconvolution is an algorithm-based process used to restore the systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.
Celleste 6 software is a powerful image analysis solution that helps make your imaging research job easier and increases your productivity by seamlessly providing images and accurate data. Coupled with the powerful image acquisition capabilities of the EVOS imaging system, you have the tools that make it easy to capture, process, measure, analyze, and share your important images and data more effectively than ever.
TRUSTED_SUSTAINABILITY
Specifications
Type Module
Product Line Celleste
For Use With (Equipment) Celleste Software (Cat. No. AMEP4942), and 2D Deconvolution Module (AMEP4864)
License Single User
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.