Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Celleste™ 2D Deconvolution Module, single-user license

Catalog No. 12563708
Change view
Click to view available options
Quantity:
1 card
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-563-708 1 card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-708 Supplier Invitrogen™ Supplier No. AMEP4864

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.

missing translation for 'validMainframeMsg'

Suite of image analysis and visualization tools designed for a range of biological applications that lets you transform your images into quantitative data in a streamlined and customizable workflow

  • Celleste 5.0 Image Analysis Software is a suite of image analysis and visualization tools designed for a range of biological applications that lets you transform your images into quantitative data in a streamlined and customizable workflow. Coupled with the powerful image acquisition capabilities of our EVOS microscopes, Celleste 5 software allows you to seamlessly capture, process, measure, analyze, and share images and data.
  • Optional modules for 2D and 3D deconvolution, visualization, and analysis allow you to customize the capabilities according to your needs. This Celleste 5 2D Blind/Non-Blind Deconvolution Module helps improve single-plane image quality (S/N) of cells or tissue slices by clearing background haze (out-of-focus light)
  • Streamlined workflow tools that make it easy to visualize and edit images
  • Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images
  • Blind and non-blind deconvolution options
  • Dramatically improved image quality
  • Removal of blur that can obscure important details
  • The tools to generate more impressive images and reliable data
  • Deconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste 5 modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Invitrogen
For Use With (Equipment) EVOS
Quantity 1 card
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.