Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Celleste™ GPU Acceleration Module for 3D Deconvolution, single-user license

Catalog No. 12563705
Change view
Click to view available options
Quantity:
1 card
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-563-705 1 card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-705 Supplier Invitrogen™ Supplier No. AMEP4876

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.

missing translation for 'validMainframeMsg'

The Celleste 3D Deconvolution GPU Acceleration Module, single user license, helps perform 3D deconvolution and analysis in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC or workstation.

The Celleste 3D Deconvolution GPU Acceleration Module, single user license, helps perform 3D deconvolution and analysis in a fraction of the time by utilizing the graphics processing unit (GPU) of your EVOS PC or workstation. It also helps enable the processing of thousands of voxels at once for faster processing speed, increased performance, and better response times. A network license including one seat is also available (Cat. No. AMEP4919). This module is used in conjunction with the Celleste 6.0 Image Analysis Software (Cat. No. AMEP4942), a full-feature image analysis suite designed for biological applications.

Celleste 6 Image Analysis with the GPU Acceleration Module offers you these important advantages:

  • Icon-based wizard-driven workflow streamlines image analysis and eliminate guesswork
  • Preconfigured and optimized analysis templates for common applications help derive the most relevant results from image sets
  • Wide range of report formats to visualize results, create reports and share images and data
  • Optional modules for 2D and 3D deconvolution, 3D visualization and 3D analysis available to meet your specific needs
  • GPU-accelerated deconvolution to increase performance and reduce the time to results
  • Clear images in a fraction of the time with CPU-based processing

More about the deconvolution
Deconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.

Celleste 6.0 software is a powerful image analysis solution that helps make your imaging research job easier and increases your productivity by seamlessly providing images and accurate data. Coupled with the powerful image acquisition capabilities of the EVOS microscopes, you now have the tools that make it easy to capture, process, measure, analyze, and share your important images and data more effectively than ever.

TRUSTED_SUSTAINABILITY

Specifications

Type Module
Compatibility EVOS™ Microscopes
For Use With (Equipment) Celleste 6.0 Image Analysis Software (Cat. No. AMEP4942)
Content And Storage Celleste 3D Deconvolution GPU Acceleration Module Card for Network License
Quantity 1 card
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.