Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform

Catalog No. cx51112
Encompass
Catalog No. CX51112 Supplier Thermo Scientific™ Supplier No. CX51112

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.

Rapidly perform high content imaging assays with this affordable, compact HCS platform.

Rapidly perform high content imaging assays with the affordable, compact Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform.

The CellInsight CX5 High Content Screening (HCS) Platform is a fast, simple-to operate, automated cellular imaging and analysis benchtop platform designed for quantitative imaging. For the researcher looking to gain the advantage of quantitative cell analysis for the first time or for those who are looking to increase scale and capacity, the CellInsight CX5 HCS Platform offers a costs effective, but powerful way to quickly automate quantitative cellular imaging.

Highlights:

  • Flexible – designed to work with or without fluorescent labeling, the new CellInsight CX5 HCS Platform uses transmitted light allowing users to explore more cell biology without the restriction of fluorescent dyes or to use the full capabilities of a five-color light engine
  • Affordable – automated high content imaging for all budgets
  • Easy to Use – quickly build and optimize image-based assays with included Thermo Scientific™HCS Studio™ 2.0 Cell Analysis Software
  • Performance – scan and analyze a 96-well plate in under four minutes
  • Quality – high-quality imaging and virtually maintenance-free operation
  • Compact – fits on a bench and is less than 2 ft. on all sides

Capabilities:

  • Make individual cell, subcellular and multicellular measurements
  • Save time by automatically collecting only the images and data you need to be statistically relevant
  • Make multiple measurements per object, over time if desired
  • Measure subpopulations and reject undesired objects automatically

Easy to Use Applications:

  • Comprehensively pre-validated and user configurable image analysis without scripting or programming
  • Exploratory assay development tools and specific biology optimized tools
  • Define biological thresholds for outputs based on control populations
  • Ability to create multi-parameter gates from individual output measurements

Hardware and Software:

  • HCS Studio 2.0 Cell Analysis Software – the premier high content instrument control, visualization and analysis program for novice to advanced users featuring image/data portability
  • Ultra-stable, long-life light source reduces variability, increases data quality, and decreases operating costs
  • Fluorescence options which include near-IR wavelengths
  • Proprietary Thermo Scientific™ AccuFocus System is fast, yet flexible, imaging cells with high-fidelity every time
  • Unique intelligent acQuisition (iQ) real time analysis feedback to optimize image collection and data quality
  • Compatible with SBS standard microplates (6-1536 well) and slides

Compatible with:
Optional Automation – Thermo Scientific™ Orbitor™ RS Plate Mover and other Thermo Scientific™ Laboratory Automation Solutions; Optional Modules - Thermo Scientific™ High Content Modules

TRUSTED_SUSTAINABILITY

Specifications

Camera Photometrics X1 CCD: High-resolution, thermoelectrically cooled to 0°C; 14-bit dynamic range; FOV 2208 x 2208 pixel array; 4.54 micron pixel size; 75% peak Quantum Efficiency (QE); USB 2.0 (no additional CameraLink or Firewire Cards needed)
Description CeLLInsight™ CX5 High Content Screening Platform with HCS Studio™ Cell Analysis Software and Store™ Image and Database Management Software
Light Source LED, solid-state five-color light engine used with provided filter sets offers the following  excitation/emission capabilities:  Ex 386/Em 440 (e.g., Hoechst); Ex 485/Em 521 (e.g., Alexa Fluor 488); Ex 560/Em 607 (e.g., TRITC); Ex 650/Em 694 (e.g., Cy5); Ex 740/Em 810 (e.g., Alexa Fluor 750)
Objectives Standard: 10X (0.30NA); 20X (0.4NA) Olympus objectives
Optional: 2x (0.08NA); 4x (0.16NA) Olympus objectives
Data Management Compatible with Thermo Scientific™ Store™Image and Database Management Software
PC Windows™ 7 64-bit PC with minimum quad core 3.7GHz processor, 8GB RAM, and 500GB Hard Drive
Software Thermo Scientific™ HCS Studio™ 2.0 Software, Thermo Scientific™ Store™ Image and Database Management Software
Depth (English) 23.4 in.
Depth (Metric) 59.4cm
Height (English) 14.6 in.
Height (Metric) 37.2cm
Weight (English) 99.2 lb.
Weight (Metric) 45kg
Width (English) 16.3 in.
Width (Metric) 41.4cm
Wattage 300w
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.