Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CellInsight™ CX7 LZR High Content Analysis Platform

Catalog No. CX7A1110LZR
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CX7A1110LZR 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CX7A1110LZR Supplier Thermo Scientific™ Supplier No. CX7A1110LZR

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.

Thermo Scientific™ CellInsight™ CX7 LZR High Content Analysis Platform is fast, laser-based, and designed for quantitative microscopy.

The CellInsight CX7 LZR HCA Platform offers a more powerful and integrated way to develop and automate high content assays using a combination of 5-color bright-field and 7-color fluorescence imaging in confocal or wide-field modes. With laser illumination, the Cell Insight LZR HCA Platform enables researchers to image faster in both wide-field and confocal mode.

Features:

  • 7-channel fluorescent imaging with laser illumination
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured to accept optional modules for fully automated plate handling and live cell on-board environmental control

Capabilities:

  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Use integrated HCS Studio software for instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Compatible with Molecular Probes fluorescent detection reagents

Hardware and software

  • High-output laser excitation light
  • Broad reagent options with 7-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software (not included) and optional Orbitor RS Microplate handler (not included)
TRUSTED_SUSTAINABILITY

Specifications

Product Line CellInsight
Color Assorted Colors
Detection Method Fluorescence
Quantity 1 each
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.