Learn More
Cellpath CellSafe 5 Core Biopsy Cassettes with Five Wells
Description
- Features integral X-ray lucent fine mesh for efficient processing
- Simultaneous preparation of core biopsies
- Core biopsies for breast and prostate samples
- Each well is numbered for identification
- Available in two colors (white and blue)
- Blue cassette produced to aid contrast and visualization of white or light colored biopsies
Specifications
Specifications
| Quantity | 100/Pk. |
| Color | White |