Learn More
Cellpath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes
Description
- Allows use of standard tissue processing cassette to clearly print 2D barcode and test infomation
- Designed using a docking area for standard processing cassette
- Strong Hex pore structure improves processing for large specimens
- Reduces turn around times
- Time saving by ensuring rapid casting of block, 30 minutes at -8°C
- Thinner chamber depths reduce specimen thickness and improves specimen quality by reducing distortion with no curling
- Better sectioning quality through adhesion of paraffin block to cassette
- Available in various colors
Specifications
Specifications
| Quantity | 500/Pk. |
| Color | Pink |