Learn More
Cellpath ProMarc System II+ Slotted Processing Cassettes, White (Loose)
Description
- Manufactured from an advanced acetal polymer blend to ensure complete solvent resistance and optimized surface properties for printing of high quality text and 2D barcodes
- Suitable for use with all widely used cassette printers including: Leica IPC, Sakura AutoWrite, Sakura Ident, Thermo Scientific PrintMate, Thermo Scientific MicroWriter,Triangle Biomedical Sciences SHUR/Mark and Vogel cassette printers
- Writing surface and angle optimised for printing of high quality text and linear/2D barcodes
- Available loose in a dispenser box
- Writing surface suitable for marking with CellMark marker pen and CellMark 2B pencil
Specifications
Specifications
| Packaging Type | Available Loose in a Dispenser Box |
| Quantity | 1000/Pk. |
| Color | White |
| Material | Acetal Polymer Blend |