Learn More
Alkali Scientific CellPro™ 1000mL Bottle Top Filter, PES Filter Material, 0.22μm, 90mm, Individually Wrapped, 24/cs
SDP
Supplier:  Alkali Scientific VH100022
Encompass_Preferred
CellPro™ Bottle Top Filters are individually wrapped and available in 0.22µm PES membrane. The screw-on funnel includes a lid and a graduated nozzle connection to fit multiple diameter vacuum hoses. They also feature engraved graduation marks to ensure accuracy and are gamma sterilized, certified RNAse, DNAse, sterile and pyrogen free. They offer extremely high flow rates at very low differential pressure. Recommended use is > -60 kPa (kilopascal) and maximum use is > -90 kPa (kilopascal). PES (Polythersulfone) material is proven to be lower protein binding than CA (Calcium Acetate) with far superior durability.
Catalog No. 50-233-6124
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.