Learn More
Alkali Scientific CellPro 15mL Centrifugal Filte
SDP
Supplier:  Alkali Scientific CF150058
Encompass
Alkali Scientific15 mL disposable ultrafiltration centrifugal devices offer a high-capacity solution for protein concentration desalting and buffer exchange of biological samples Designed with a 50 kDa nominal molecular weight limit (NMWL) membrane these devices enable fast and efficient sample processing while achieving over 90% recovery Ideal for larger-volume applications the device accommodates a maximum initial volume of 15 mL and concentrates samples down to a typical final volume of 200 L Its durable single-use polypropylene construction ensures chemical resistance and sample integrity while compatibility with both swinging-bucket and fixed-angle rotors provides flexibility With a maximum RCF of 3000 g for swinging-bucket rotors and 4000 g for fixed-angle rotors these devices deliver efficient and reliable ultrafiltration performance Material Composition Outer Tube Polypropylene (PP) Filter Methacrylate-butadiene-styrene (MBS) Filter Membrane Polyethersulfone (PES) Tube
Catalog No. 50-302-6549