Learn More
Alkali Scientific CellPro .5mL Centrifugal Filte
SDP
Supplier:  Alkali Scientific CF0500525
Encompass
Alkali Scientific0 5 mL disposable ultrafiltration centrifugal devices provide a highly efficient solution for protein concentration desalting and buffer exchange of biological samples Equipped with a 50 kDa nominal molecular weight limit (NMWL) membrane these devices enable rapid sample processing while achieving over 90% recovery Designed for small-volume applications the device supports a maximum initial volume of 0 5 mL and concentrates samples down to a final volume of approximately 510 L Its single-use polypropylene construction ensures durability and chemical resistance while compatibility with both swinging-bucket and fixed-angle rotors allows for versatile use With a maximum RCF of 14000 g these devices offer fast and reliable ultrafiltration performance Material Composition Outer Tube Polypropylene (PP) Filter Methacrylate-butadiene-styrene (MBS) Filter Membrane Polyethersulfone (PES) Tube Cap High-density polyethylene (HDPE) Key Features 50 kDa NMWL membrane for effic
Catalog No. 50-302-6409