Learn More
Alkali Scientific CellPro™ DEPC Ultra Pure Water, 20L Cubitainer
SDP
Supplier:  Alkali Scientific DC4220L
Encompass
CellPro™ DEPC treated Ultra Pure Water is specially treated with diethyl pyrocarbonate (DEPC) to inactivate ribonucleases (RNases), ensuring the purity of the water for molecular biology applications. DEPC treatment effectively eliminates RNase contamination, making this ultra-pure water ideal for use in RNA-related experiments, such as RNA isolation, reverse transcription, and RNA sequencing.
Catalog No. 50-233-5875