Learn More
Alkali Scientific ASI™ Laboratory Label, Color Coded Dot – Sheet Form - 1", 42 Sheets, Pink Flr
SDP
Supplier:  Alkali Scientific DOT10M PINK FLR
Encompass_Preferred
Label, Color Coded Dot – Sheet Form - 1" We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).
Catalog No. 50-302-6636
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.