Learn More
Alkali Scientific CellPro™ DMEM/F12 1:1 medium, with HEPES; without L-glutamine, 500ml, 10/Cas
Supplier: Alkali Scientific F12005CS
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.