Learn More
Alkali Scientific ASI™ Fetal Bovine Serum (FBS), Australian Source, USDA Approved, 500mL
Supplier: Alkali Scientific FB75
CellPro™ Fetal Bovine Serum raw material is sourced from USDA approved abattoirs in Australia and imported to the US for processing and final filtration. Material undergoes USDA quarantine and safety testing prior to production. Virus-screened Mycoplasma-tested Triple 0.1 Micron Filtered Low Hemoglobin lots (<15mg/dL) may be available. Low Endotoxin lots (<1.0EU/mL) may be available. This product ships on dry ice.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.