Learn More
Alkali Scientific CellPro™ Pestles for FlowTubes with 35um Strainer Caps, Sterile, Individually Wrapped 100/pk
SDP
Supplier:  Alkali Scientific CT6045
Encompass_Preferred
Pestles For FlowTube™ Culture Tubes For BD FACS Designed will homogenize the sample perfectly through the 35µm mesh cap Compatible with FACS (BD) instruments Blue Polystyrene Sterile; RNase & DNase free
Catalog No. 50-302-9151
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.