Learn More
Alkali Scientific BioBucket™ collapsible laboratory bucket, 5 Liter, 1/each
SDP
Supplier:  Alkali Scientific R8300
Encompass
BioBucket™ collapsible laboratory bucket, 5 Liter, 1/each is ageneral laboratory container that every lab could benefit from. Its collapsible nature makes it the ideal container for any lab looking to conserve storage space. BioBucket can be used as an ice bucket to keep samples cool over long periods of time, or as portable liquid container. The attached handle makes transportation a breeze while the molded base grip allows for easy pouring and cleaning. The BioBucket is perfect for field use as an environmental sample collection and transport receptacle.  Collapsible body saves valuable storage space Useable with crushed ice Inner-lid access port for enhanced temperature retention Attached handle for easy transportation Molded grip on base for pouring
Catalog No. 50-302-8953