Learn More
Alkali Scientific CellPro Syringe Filter, PES 33mm, 0.45um, STERILE, Individually Wrapped, 50/Pk
SDP
Supplier:  Alkali Scientific TE34S
Encompass_Preferred
These Syringe Filters consists of a plastic housing with a membrane which serves as a filter for fluids. The filter is designed to press fluid through the syringe filter.Specifications:• Ultrasonic Welded • 25mm Filter Diameter for volumes between 10-100mL• Filter Inlet: Luer Lock Female, Filter Outlet: Luer Male• Maximum operating pressure for 25mm is 7Bar, recommended to use syringe sizes of 3mL or larger• Pore Size of 0.20 µm optimal for sterile filtration or 0.45 µm optimal for thorough cleaning• PVDF, Polyvinylideneflouride Characteristics:Hydrophilic membrane with good solvent resistance. Low UV absorbing extractables and low non-specific binding Applications: PVDF is used for general biological filtration, and filtration of samples where high protein recovery is desired. It has slight hydrophilic properties and can be used for clear filtration and sterilization of aqueous solutions• PES, Polyethersulfone Characteristics:High flow rates with good throug
Catalog No. 50-233-6175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.