Learn More
Thermo Scientific™ CellSensor™ AP-1-bla A375 Cell Line
Description
The CellSensor™ AP1-bla A375 cell line contains a beta-lactamase reporter gene under control of the AP1 response element stably integrated into A375 cells. A375 cells are human melanoma cancer cells that contain the endogenous B-Raf mutation V600E resulting in constitutive B-Raf kinase activity. The CellSensor™ AP1-bla A375 cell line is a clonal population isolated by flow cytometry based on constitutive expression of beta-lactamase.
This cell line has been validated with various small-molecule Raf inhibitors as well as B-Raf Stealth RNAi™ siRNA. This cell line has also been tested for assay performance under variable conditions, including DMSO concentration, cell number, compound incubation time, and substrate loading time and validated for Zfi-factor and IC50 concentrations of Raf1 Inhibitor I. The RAF gene family (RAF1, A-RAF, and B-RAF) encodes closely related serine/threonine protein kinases that are important effectors of Ras activation. Raf1 and A-Raf are rarely mutated, whereas mutations in B-Raf are common in human cancers, especially melanoma. B-Raf is mutated in about 70% of human melanomas, 35-70% of papillary thyroid carcinomas, and less commonly in lung and colorectal carcinomas. Mutations are mostly in the B-Raf kinase domain and, in melanomas, the vast majority are V600E missense mutations leading to activation of B-Raf kinase. The constitutive activity of B-Raf V600E can directly lead to the activation of the Mek/MAPK signaling pathway. Therefore, inhibition of B-Raf/Mek/MAPK signaling could be a potential way of treating melanomas and other tumors with mutant B-Raf.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | MAPK pathway, BRAF V600E |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.