Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellSensor™ ARE-bla Hep G2 Cell Line

Catalog No. K1633
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1633 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1633 Supplier Invitrogen™ Supplier No. K1633
Season of Savings
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CellSensor™ ARE-bla Hep G2 cell line

The CellSensor™ ARE-bla Hep G2 cell line is responsive to tBHQ and Sulforaphane and can be used for analyzing the Nrf2/antioxidant response signaling pathway using the GeneBLAzer™ Loading Substrates. Reactive oxygen species (ROS) can damage biological macromolecules and are detrimental to cellular health. Electrophilic compounds, xenobiotics and antioxidants are sources of reactive oxygen species, creating oxidative stress that can harm cells. Enzymes are involved in the Phase II detoxification of xenobiotics to reduce cellular stress include glutathione transferases, quinone reductase, epoxide hydrolase, heme oxygenase, UDP-glucuronosyl transferases, and gamma-glutamylcysteine synthetase. Expression of these genes protects cells from oxidative damage and can prevent mutagenesis and cancer. Transcription of these enzymes is coordinately regulated through antioxidant response elements (AREs). Nrf2 (NF-E2-related factor 2) and Nrf1 are transcription factors that bind to AREs and activate these genes. The CellSensor™ ARE-bla Hep G2 cell line contains a beta-lactamase reporter gene under control of the Antioxidant Response Element (ARE) stably integrated into Hep G2 cells. To construct this cell line, the ARE-bla construct was transduced into Hep G2 cells. Flow cytometry was used to isolate cells responsive to tert-butylhydroquinone (tBHQ). This cell line is tested under various experimental conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, and functionally validated for Z'; and EC50 concentrations of tBHQ. Academic and non-profit customers, please inquire for special pricing.
TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Product Line CellSensor
Quantity 1 vial
Target Entry Anti-oxidant response pathway, NRF2
Type Cell Line
Cell Line Hep G2
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.