Learn More
Thermo Scientific™ CellSensor™ CRE HEK 293T DA Assay Kit
Catalog No. K1457
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
Catalog No. Quantity
K1457 1 Plate
1 options
Catalog No. K1457 Supplier Thermo Scientific™ Supplier No. K1457
The CellSensor™ CRE HEK 293T DA (Division Arrested) cells contains a beta-lactamase (bla) reporter gene under control of a cAMP response element (CRE) stably integrated into HEK293T cells.
The CellSensor™ CRE HEK 293T DA (Division Arrested) cells contains a beta-lactamase (bla) reporter gene under control of a cAMP response element (CRE) stably integrated into HEK293T cells. Fluorescence activated cell sorting (FACS) was used to isolate clones responsive to stimulation of the CRE pathway by forskolin. The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading conditions, and EC50 concentration of forskolin. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
This product can serve as a negative control in screening assays performed with specific G-protein coupled receptor (GPCR) DA cells that were made from this same background. These cells can be used in other assays where a sensitive readout to intracellular changes to cAMP is needed.
TRUSTED_SUSTAINABILITY
Specifications
Cell State Division-Arrested Cells
Content And Storage Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
Includes:
• CellSensor™ CRE HEK293T DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D
• CellSensor™ CRE HEK293T CHO-K1 Assay Protocol
• LiveBLAzer™-FRET B/G Loading Protocol
• Certificate of Analysis
The CellSensor™ CRE HEK 293T DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Product Line CellSensor
Quantity 1 Plate
System Type CellSensor™
Druggable Target Signaling Pathway
For Research Use Only. Not for use in diagnostic procedures.