Learn More
Thermo Scientific™ CellSensor™ CRE-bla CHO K1 Cell Line
Description
The CellSensor™ CRE-bla CHO-K1 cell line contains a beta-lactamase reporter gene under control of a cAMP response element (CRE) stably integrated into CHO-K1 cells. This cell line can be used as a parental cell line to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest or to detect changes in intracellular cAMP levels. CRE-bla CHO-K1 cells have been shown to be responsive to forskolin stimulation. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | PKA, cAMP, cAMP/PKA pathway |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.