Learn More
Thermo Scientific™ CellSensor™ IL1-bla ECV304 Cell Line
Description
The CellSensor™ IL-1-bla ECV304 cell line is an adherent human urinary bladder cell line used to measure the (IL-1) signaling pathway via transcriptional regulation of the beta-lactamase gene. IL-1 cytokine cell signaling is a critical pathway involved in immune and inflammatory responses. This cell line was constructed using GenomeScreen™ technology utilizing a promoterless beta-lactamase reporter gene randomly integrated into the genome of the IL-1-bla ECV304 cell line. Sequential rounds of sorting were performed to isolate cell clones containing tagged genes induced by IL-1. A dose response assay was run with this cell line using IL-1 as the agonist. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | Interleukin 1 pathway, IRAK |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.