Learn More
Thermo Scientific™ CellSensor™ irf1-bla TF-1 Cell Line
Description
Jak/Stat signaling pathways play essential roles in cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin-3 (IL-3), prolactin, erythropoietin (Epo), and granulocyte-macrophage colony stimulating factor (GM-CSF). JAK2 gene knockout causes embryonic lethality due to defective erythropoiesis, suggesting that the Jak2/Stat5 pathway plays an important role in red blood cell formation.
The recent discovery of an activating mutation in JAK2 (V617F) present in a high percentage of myeloproliferative disease (MPD) patients suggests that the Jak2/Stat5 pathway may be a potential therapeutic target for certain forms of MPD. The activated Stat5 transcription factor recognizes and binds to a specific palindromic DNA sequence found in the promoter region of β-casein, interferon regulatory factor-1 (irf-1), and a number of other genes. The CellSensor™ irf1-bla TF1 Cell Line contains a beta-lactamase reporter gene under control of the irf-1 response element stably integrated into TF1 cells. TF1 cells are a human erythroleukemia cell line that is growth-dependent on GM-CSF and have an intact GM-CSF-JAK2-STAT5 pathway.
This cell line is validated for EC50 and Z'-factor using GM-CSF. This cell line has also been tested under variable experimental conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Responsiveness to EPO and Jak Inhibitor 1 was also tested (Figures 2 and 3). CellSensor™ irf1-bla TF1 cells were stimulated in triplicate with GM-CSF over the indicated concentration range in a 384-well format. Cells were incubated for 5 hr. with agonist and 0.5% DMSO and then combined with LiveBLAzer™-FRET B/G Substrate (CCF4-AM) for 2.5 hr. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The 460/530 ratios were plotted for each replicate against the indicated concentrations of GM-CSF. CellSensor™ irf1-bla TF1 cells were stimulated with Epo over the indicated concentration range in a 384-well format.
The Response Ratios were plotted against the indicated concentrations of Epo. CellSensor™ irf1-bla TF1 cells were treated with Jak Inhibitor 1 over the indicated concentration range in a 384-well format. Cells were then stimulated with GM-CSF or EPO for 5 hr. in 0.5% DMSO and then combined with LiveBLAzer™-FRET B/G Substrate (CCF4 - AM) for 2.5 hr. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. These values were converted to percent inhibition relative to a set of controls (unstimulated cells and EC80 of GM-CSF or EPO-treated cells) and plotted against the indicated concentrations of Jak Inhibitor 1.
Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | JAK/STAT (GM-CSF, EPO, IL-3) pathway, JAK2, STAT5 |
| Type | Cell Line |
| Cell Line | TF-1 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.