Learn More
Thermo Scientific™ CellSensor™ ISRE-bla HEK293T Cell Line
Description
Interferon cell signaling is a critical pathway involved in viral defense and autoimmune disease. The CellSensor™ ISRE-bla HEK 293T cell line is responsive to interferon alpha and can be used to probe the JAK/STAT signaling pathway. The CellSensor™ ISRE-bla HEK 293T cell line contains the β-lactamase reporter gene under the control of the interferon stimulated response element (ISRE). This cell line was engineered by transduction of the ISRE-bla construct into HEK 293T cells by lentivirus. Flow cytometry was then used to isolate a pool of clones responsive to interferon alpha a stimulation. A dose response curve for this assay against interferon alpha is available in the validation packet. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | JAK/STAT (Type I IFN) pathway, TYK2, JAK1 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.