Learn More
Thermo Scientific™ CellSensor™ NFAT bla HEK 293T Cell Line
Description
The CellSensor™ NFAT-bla HEK 293T cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into HEK 293T cells. Fluorescence activated cell sorting (FACS) was used to isolate clones responsive to stimulation of the NFAT pathway by PMA (Phorbol 12-Myristate 13-Acetate) and Thapsagargin (Thaps). The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading conditions, and EC50 concentration of Thaps at fixed concentration of PMA. The CellSensor™ NFAT-bla HEK 293T cell line responds to agonist treatment as expected from the literature and can be adapted for high-throughput screening (including coupling of receptors to the NFAT pathway) for agonists or antagonists with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | PKC, PKC and calcium pathway |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.