Learn More
Thermo Scientific™ CellSensor™ NFAT CHO-K1 DA Assay Kit
Description
The CellSensor™ NFAT CHO-K1 DA (Division Arrested) cells contain a beta-lactamase (bla) reporter gene under control of a Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into CHO-K1 cells. Fluorescence activated cell sorting (FACS) was used to isolate clones responsive to stimulation of the NFAT pathway by PMA (Phorbol 12-Myristate 13-Acetate) and Thapsagargin (Thaps). The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading conditions, and EC50 concentration of Thaps at a fixed concentration of PMA. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction
This product can serve as a negative control in screening assays performed with specific G-protein coupled receptor (GPCR) DA cells that were made from this same background. These cells can also be used in other assays where a sensitive readout to intracellular changes in calcium is needed.
Specifications
Specifications
| Cell State | Division-Arrested Cells |
| Content And Storage | Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol). Includes: • CellSensor™ NFAT CHO-K1 DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D • CellSensor™ NFAT CHO-K1 Assay Protocol • LiveBLAzer™-FRET B/G Loading Protocol • Certificate of Analysis The CellSensor™ NFAT CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Product Line | CellSensor |
| Quantity | 1 Plate |
| System Type | GeneBLAzer™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.