Learn More
Thermo Scientific™ CellSensor™ STAT6-bla RA-1 Cell Line
Description
The CellSensor™ STAT6-bla RA-1 Cell Line contains a beta-lactamase reporter gene under control of the STAT6 response element stably integrated into Ramos-1 (RA-1) cells. This cell line is a clonal population isolated by flow cytometry in response to interleukin-4 (IL-4). This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time, and validated for Zfi-factor and EC50 concentrations of IL-4.
Additional data using alternate stimuli are also available. IL-4 is known to induce the growth and differentiation of B cells, T cells, myeloid cells, and hepatocytes. In B cells, IL-4 acts as a co-mitogen to induce the expression of the Fc receptor for IgE and also major histocompatibility complex (MHC) class II molecules, and it stimulates the transcription of immunoglobulin heavy-chain germline IgE and IgG1 genes, leading to class switching. IL-4 function is mediated by the IL-4 receptor complex, which activates Jak1 and Jak3 tyrosine kinases. STAT6 is recruited to the IL-4R complex and is subsequently phosphorylated by the Jak kinases. This causes STAT6 to dimerize and translocate to the nucleus where it binds to specific sequences on IL-4-responsive genes. Optimal transcription response of IL-4-inducible promoters requires two Th2-mediated stimuli, CD40 ligand and IL-4. CD40 ligand is a protein expressed on the surface of T cells. It regulates B cell function by engaging CD40 on the B cell surface, leading to signaling through NFκB. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Target Entry | STAT6, Interleukin 4/STAT6 pathway, JAK3 |
| Type | Cell Line |
| Cell Line | RA-1 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.