Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellTrace™ Calcein Blue, AM - Special Packaging

Catalog No. 501137412
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7412 20 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-7412 Supplier Invitrogen™ Supplier No. C34853
  • $508.00
    $380.65 / Each of 1

    Save $127.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

CellTrace™ calcein blue, AM - special packaging

CellTrace calcein blue AM is a proven short-term, blue-fluorescent tracer for cell labeling. The probe possesses acetoxymethyl (AM) esters that allow its passive diffusion across cell membranes. Upon cleavage of the AM esters by intracellular esterases, the molecule that remains (absorption/emission maxima ∼360/449 nm) is relatively polar and is retained by cells for several hours.

Related Links:

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 20 vials of calcein blue (50 μg per vial). Store in freezer (-5 to -30°C).
Excitation Wavelength Range 323⁄439
Dye Type calcein blue
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Quantity 20 x 50 μg
Detection Method Fluorescence
Emission UV
Form Solid
Product Line CellTrace
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Label Type Other Labels or Dyes
Product Type Viability Indicator
Show More Show Less
I need a general cytoplasmic stain that does not overlap with the GFP in my cells. What do you recommend?

Calcein AM, a green dye, is typically used as a general cytoplasmic stain, but not recommended with GFP-positive cells. For GFP-expressing cells there are other colors available: Calcein Blue AM, Calcein Violet AM, and Calcein Red-Orange AM. The retention time of these dyes in live cells is dependent upon the inherent properties of the cell.

I stained two populations of cells, one with CellTracker Green and the other with CellTracker Red, but it looks like there may be crossover of the red dye to the green cells. What is going on?

One possibility is that there is spectral bleedthrough between the dyes. Be sure to check the single-color samples by imaging the red cells in green and imaging the green cells in red, using the optimal imaging settings for the other color. If you see bleedthrough with these controls, then you will have to reduce the dye label concentration to reduce the brightness of the dyes, or choose dyes that are farther apart spectrally. If the issue isn’t bleedthrough, another possibility is that the cells were not adequately washed after staining, allowing some unincorporated dye to remain and label the other cells after they were introduced. Extending washes and wash times should help with this.

I stained my cells with Calcein, AM, but the signal went away after I fixed my cells. Why is this?

Calcein, AM diffuses into cells, the 'AM' moiety is cleaved by cellular esterases, and then the dye molecules are observed in the cytoplasm without binding to anything. This gives a 'whole cell' stain. It also means that the dyes are not crosslinked with aldehyde-based fixation and thus will be lost upon fixation. Additionally, any disruption of plasma membrane, such as with detergents or trypsinization, will lead to leakage of the dye from the cell.

I'm trying to stain my cells with CellTracker dyes or CFDA SE, but I'm not seeing much signal. What can I do?

First, make sure you aren’t staining in the presence of serum, since serum can have esterase activity that can prematurely cleave the AM group on these dyes, preventing entry into cells. After staining, it’s okay to return the cells to medium containing serum. After this, you can try increasing the concentration and label time to get a higher intensity.

I like how calcein dyes label the whole cell. How long can I track my cells with them, and can I fix them?

Calcein dyes diffuse into cells, the 'AM' moiety is cleaved by cellular esterases and then are observed in the cytoplasm without binding to anything. This provides a 'whole cell' label. Calcein dyes may be pumped out by normal cellular efflux mechanisms, sometimes within a very short time, especially for cell types that may exhibit drug resistance, unless the efflux is inhibited (such as with probenecid). The dyes are not crosslinked with aldehyde-based fixation, unlike protein-binding CellTracker dyes, and thus will be lost upon fixation. Additionally, any disruption of plasma membrane, such as with detergents or trypsinization, will lead to leakage of the dyes from the cell.

I am trying to assay cell proliferation with a CellTrace stain, and I am not seeing separate peaks for each cell division. How can I optimize this assay?

The key to good generational profiles with CellTrace reagents is starting with cells that are evenly labeled so that they have a tight coefficient of variance (CV) when run at time zero after labeling. If the peak is too broad, the generations will overlap each other and the series of peaks will become a hump. Even labeling can be achieved by starting with a uniform cell population (not a mixture of lymphocytes and granulocytes for example) as staining will be proportional to cell size. Cells are labeled rapidly, so you want to pre-dilute the dye and mix it into your cells rapidly. Be sure that the cells are not sitting in a clump in the bottom of your tube. The easiest way to do this is to make a 2x dye solution (1x = 1-10 µM) and resuspend your cells in a half volume of medium (no serum or BSA). Add the dye to the cells and invert a few times to mix. Gently agitate the cells during staining. Once the dye incubation is over (20 min, 37 degrees C), add serum or BSA (at least 1%) to scavenge any remaining unreacted dye. Spin down cells, wash 1x, and resuspend in complete medium. After a 10-20 min incubation to undergo de-esterification, cells are ready to be set up for whatever treatment you are planning. Be sure to keep a time zero control as you need to know where the first generation ran.

What kinds of cell health and viability assays can be performed by flow cytometry?

The following cell health and viability assays can be performed by flow cytometry :

-Apoptosis Assays:
Membrane Asymmetry: Annexin V is a member of a family of structurally related proteins that bind phospholipids in the presence of Ca2+. Annexin V binds several phospholipids, but shows highest affinity for phosphatidylserine.
Phosphatidylserine is normally found in the inner leaflet of the cell membrane; however, in the early stages of apoptosis, phosphatidylserine is observed to translocate to the outer leaflet. This translocation makes phosphatidylserine available for annexin V binding in the presence of Ca2+ containing incubation buffer. Cells undergoing apoptosis will stain with annexin V, while normal cells will not. annexin V is available conjugated with a wide range of fluorophores.

Mitochondrial Health: A distinctive feature of the early stages of apoptosis is the disruption of the mitochondria, including changes in membrane and redox potential. We exclusively offer a number of fluorescent probes for analyzing mitochondrial activity in live cells by flow cytometry, with minimal disruption of cellular function.

The MitoProbe family of mitochondrial stains (MitoProbe DiOC2(3) Assay Kit, Cat. No. M34150, MitoProbe JC-1 Assay Kit, Cat. No. M34152, and MitoProbe DiIC1(5) Assay Kit, Cat. No. M34151) provides quick, easy, and reliable flow cytometric detection of the loss of mitochondrial membrane potential that occurs during apoptosis.

Caspase Activity: The CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit (Cat. No. C10427) enables flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells. The kit includes the novel fluorogenic substrate CellEvent Caspase-3/7 Green Detection Reagent which targets the recognition sequence for activated caspase-3 and caspase-7, as well as SYTOX AADvanced Dead Cell Stain.

DNA Fragmentation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. DNA fragmentation that occurs during apoptosis produces DNA strand breaks, and can be analyzed using TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) assays. The APO-BrdU TUNEL assay (Cat. No. A23210) is a two-color assay for labeling DNA breaks and total cellular DNA to detect apoptotic cells by imaging or flow cytometry.

Nuclear Chromatin Condensation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. Cells undergoing apoptosis display an increase in nuclear chromatin condensation. As the chromatin condenses, cell-permeable nucleic acid stains becomes hyperfluorescent, thus enabling the identification of apoptotic cells when combined with a traditional dead-cell stain. The Vybrant Apoptosis Assay Kit #5, Hoechst 33342/Propidium Iodide (Cat. No. V13244) provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. The Chromatin Condensation & Membrane Permeability Dead Cell Apoptosis Kit with Hoechst 33342, YO-PRO-1, and PI dyes, for flow cytometry (Cat. No. V23201) detects apoptotic cells with changes in nuclear chromatin condensation and plasma membrane permeability.

-Cell Cycle Analysis:
Live cell assays: The Vybrant DyeCycle family of dyes offers robust fluorescent dyes for live-cell cycle analysis with limited cytotoxicity using 405 nm (Cat. No. V35003), 488 nm (Cat. No. V35004), 532 nm (Cat. No. V35005), or 633 nm (Cat. Nos. V10309 and V10273) excitation. The dyes have low cytotoxicity, allowing stained cells to be sorted and otherwise cultured or assessed with functional assays after staining.

Fixed cell assays: Analyzing cell cycle using FxCycle Violet Stain (Cat. No. F10347), SYTOX AADvanced Dead Cell Stain Kit (Cat. No. S10349) or FxCycle Far Red Stain (Cat. No. F10348) allows for multiple color options for simplified fixed cell cycle analysis.

-Cell Proliferation:
Dye dilution assays for cell proliferation: Dye dilution assays for cell proliferation rely on cell membrane–permeant fluorescent molecules. Upon entry into the cell, the dye will covalently bind to amine groups on proteins, resulting in long-term dye retention within the cell. Through subsequent cell divisions, each daughter cell receives approximately half the fluorescence of the parent. Analysis of the fluorescence intensities of cell populations by flow cytometry enables determination of the number of generations through which a cell or population has progressed since the label was applied. CellTrace fluorescent stains can be used without affecting morphology or physiology to trace generations in vivo or in vitro. There is no known effect on proliferative ability or biology of cells and they are well retained in cells for several days post-stain. Available kits for flow cytometry include CellTrace CFSE Cell Proliferation Kit (Cat. No. C34554), CellTrace Violet Cell Proliferation Kit (Cat. No. C34557), and CellTrace Far Red Cell Proliferation Kit (Cat. No. C34564).

DNA Synthesis Assays: Measuring the synthesis of new DNA is a precise way to assay cell proliferation in individual cells or in cell populations. DNA synthesis–based cell proliferation assays measure the rate of new DNA synthesis based on incorporation of modified nucleosides. The Click-iT Plus EdU cell proliferation assay utilizes the power of click chemistry and the modified nucleoside EdU to provide a superior alternative to BrdU staining for detecting and quantitating newly synthesized DNA. The Click-iT Plus EdU cell proliferation assay is available with Pacific Blue (Cat. No. C10636), Alexa Fluor 488 (Cat. Nos. C10632 and C10633), and Alexa Fluor 647 (Cat. Nos. C10634 and C10635).

-Viability Assays:
Dead cells often give false positive results, as they tend to bind non-specifically to many reagents. Therefore, removing dead cells from your flow cytometry data is a critical step to help ensure accurate results and analysis.

Non-fixable Membrane Permeability Stains: SYTOX Dead Cell Stains (Cat. Nos. S34857, S34860, S34861, S34859, and S34862) do not cross intact cell membranes, and they exhibit increased fluorescence upon dsDNA binding, making them some of our most brilliant dead cell stains. Cell-impermeant classic DNA-binding dyes include propidium iodide (Cat. No. P21493) and 7-AAD (Cat. No. A1310). Both of these dyes have been used extensively for viability assays in flow cytometry. CellTrace Calcein AM dyes can be passively loaded into adherent and nonadherent cells. These cell-permeant esterase substrates serve as viability probes that measure both enzymatic activity, which is required to activate their fluorescence, and cell membrane integrity, which is required for intracellular retention of their fluorescent products. Available with blue (Cat. No. C34853), violet (Cat. No. C34858), and green (Cat. No. C34852) fluorescence, these dyes are ideal for short-term staining of live cells and can be used in multiplexed flow cytometry experiments.

Fixable Viability Stains: The LIVE/DEAD Fixable Dead Cell Stains are fixable viability dyes that help to ensure accurate assessment of cell viability in samples after fixation and/or permeabilization. LIVE/DEAD Fixable Dead Cell Stain Kits are based on the reaction of a fluorescent reactive dye with cellular proteins (amines). These dyes cannot penetrate live-cell membranes, so only cell-surface proteins are available to react with the dye, resulting in dim staining. The reactive dye can permeate the damaged membranes of dead cells and stain both the interior and exterior amines, resulting in more intense staining. LIVE/DEAD Fixable Dead Cell Stain Kits are available in eight single-channel colors available for UV, 405, 488, 532, 561, or 633 nm lasers in three packaging sizes to match your experiment.

What are the advantages of flow cytometry?

-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.

What kinds of applications can I run on a flow cytometer?

There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.