Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Spunbond Polypropylene Lab Coats, Elastic Wrists

Catalog No. 19304133 Shop All Cellucap Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-304-133 Large
19-304-131 Small
19-304-136 3X-Large
19-304-134 X-Large
19-304-132 Medium
19-304-135 2X-Large
19-304-139 6X-Large
19-304-138 5X-Large
19-304-140 7X-Large
19-304-137 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-304-133 Supplier Cellucap Supplier No. 3302EWHLL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-woven material lab coats offer lightweight splash protection from non-hazardous liquids and dry particulates.

  • Lab coat offers lightweight splash protection from non-hazardous liquids and dry particulates
  • 1.25 oz spunbond polypropylene
  • Snap front
  • Packaged 25 per case
  • No pockets, elastic wrists

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Model 3302EW
For Use With (Application) Splash protection from nonhazardous liquids and dry particulates
Item Description Velcro™
Weight (English) 1.25 oz.
Weight (Metric) 0.03 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.