Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Skid-Resistant Spunbond Polypropylene Shoe Covers

Catalog No. 19088128 Shop All Cellucap Products
Change view
Click to view available options
Size:
Universal
X-Large
Packaging Type:
Bags
Dispenser Boxes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19-088-128 Universal Bags
M28033 X-Large Dispenser Boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-088-128 Supplier Cellucap Supplier No. 2803B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Skid-resistant shoe covers provide lightweight protection from non-hazardous liquids and dry particulates. Ideal for visitors or light duty applications.

  • Sponbond polypropylene
  • Non-conductive
  • Lightweight
  • 150 pair per case, bags or boxes

Specifications

Size Universal
Material Spunbond Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Sole Type Non-skid
Model 2803B
For Use With (Application) Visitors, Light Duty, General Purpose Maintenance, Cleaning
Packaging Quantity 150 Pairs/Cs.
Packaging Type Bags
Resistance Fluid
Product Type Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.