Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cellular Antioxidant Assay Kit, Fluorescent, Adherent Cells, 192 Tests

Catalog No. 502585938
Encompass_Preferred
Change view
Click to view available options
Quantity:
192 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5938 192 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5938 Supplier Abcam Supplier No. AB242300192TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fluorescent assay kit for measuring antioxidant activity in adherent cells.

  • Detection method: Fluorescent
  • Sample types: Adherent cells
  • Cited in: 1 publication

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Benzoic acid, 2-[3,6-bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]-
Substrate Properties Chemical Substrate
Substrate Benzoic acid, 2-[3,6-bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]-
No. of Reactions 192 Tests
Product Type Antioxidant
Detection Location In-Cell Detection
Format Kit
Detection Method Fluorescence
Sample Type Adherent cells
Quantity 192 Tests
For Use With (Application) Cellular antioxidant assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.